Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033606.2 | NP_001028778.2 | 6 - 7 | 1271 | 139 | |
NM_001136222.1 | NP_001129694.1 | 5 - 6 | 853 | 139 | ||
NM_028817.3 | NP_083093.2 | 7 - 8 | 1411 | 139 | ||
GenBank mRNA | AK012088.1 | - | 6 - 7 | 1214 | 139 | |
AK160839.1 | - | 5 - 6 | 797 | 139 | ||
AK161708.1 | - | 7 - 8 | 1412 | 139 | ||
AK166723.1 | - | 7 - 8 | 1349 | 139 | ||
BC031529.1 | - | 5 - 6 | 996 | 4009004 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |