Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026319.3 | NP_080595.2 | 16 - 17 | 1514 | 110 | |
GenBank mRNA | AK006492.1 | - | 7 - 8 | 621 | 110 | |
BY706530.1 | - | 7 - 8 | 619 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |