Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172695.2 | NP_766283.2 | 2 - 3 | 562 | 114 | |
GenBank mRNA | AK019070.1 | - | 2 - 3 | 567 | 114 | |
AK075934.1 | - | 2 - 3 | 494 | 114 | ||
AK081641.1 | - | 2 - 3 | 452 | 114 | ||
AK166122.1 | - | 2 - 3 | 467 | 114 | ||
BC139355.1 | - | 2 - 3 | 561 | 8862377 | 114 | |
BC139356.1 | - | 2 - 3 | 561 | 8862378 | 114 | |
BC139773.1 | - | 2 - 3 | 472 | 40111789 | 114 | |
CA490654.1 | - | 2 - 3 | 599 | 113 | ||
M57958.1 | - | 424 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |