Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cachd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198037.1 | NP_932154.1 | 21 - 22 | 3467 | 65 | |
GenBank mRNA | AK046646.1 | - | 3382 | 65 | ||
BC057356.1 | - | 1753 | 65 | |||
BC058676.1 | - | 21 - 22 | 3467 | 6834428 | 65 | |
BC080313.1 | - | 1751 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |