Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110202.1 | NP_001103672.1 | 9 - 10 | 2113 | 129 | |
NM_001286387.1 | NP_001273316.1 | 11 - 12 | 2332 | 129 | ||
NM_001286388.1 | NP_001273317.1 | 11 - 12 | 2335 | 129 | ||
NM_053167.3 | NP_444397.2 | 11 - 12 | 2335 | 129 | ||
GenBank mRNA | AK028280.1 | - | 9 - 10 | 1832 | 129 | |
AK049907.1 | - | 11 - 12 | 2049 | 129 | ||
AK129109.1 | - | 11 - 12 | 2040 | 129 | ||
AK163123.1 | - | 11 - 12 | 2069 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |