Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286387.1 | NP_001273316.1 | 9 - 10 | 2265 | 78 | |
NM_001286388.1 | NP_001273317.1 | 9 - 10 | 2268 | 78 | ||
NM_053167.3 | NP_444397.2 | 9 - 10 | 2268 | 78 | ||
GenBank mRNA | AK049907.1 | - | 9 - 10 | 1982 | 78 | |
AK129109.1 | - | 9 - 10 | 1973 | 78 | ||
AK163123.1 | - | 9 - 10 | 2002 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |