Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ythdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163013.1 | NP_001156485.1 | 6 - 7 | 1012 | 148 | |
GenBank mRNA | BC072654.1 | - | 1 - 2 | 58 | 148 | |
BC138263.1 | - | 6 - 7 | 1012 | 8861282 | 148 | |
BC171951.1 | - | 6 - 7 | 1012 | 9054237 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |