Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acadl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007381.4 | NP_031407.2 | 3 - 4 | 428 | 82 | |
GenBank mRNA | AK005140.1 | - | 3 - 4 | 387 | 82 | |
AK018319.1 | - | 2 - 3 | 204 | 82 | ||
AK151845.1 | - | 3 - 4 | 397 | 82 | ||
AK167537.1 | - | 3 - 4 | 397 | 82 | ||
BC027412.1 | - | 3 - 4 | 385 | 4236225 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |