Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019781.2 | NP_062755.1 | 1 - 2 | 48 | 62 | |
GenBank mRNA | AF097512.1 | - | 1 - 2 | 48 | 62 | |
AK148779.1 | - | 1 - 2 | 52 | 62 | ||
BC028952.1 | - | 1 - 2 | 45 | 5368312 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |