Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golph3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177670.1 | NP_001171141.1 | 1 - 2 | 196 | 85 | |
NM_146133.3 | NP_666245.2 | 1 - 2 | 196 | 85 | ||
GenBank mRNA | AK008434.1 | - | 1 - 2 | 163 | 85 | |
AK167675.1 | - | 1 - 2 | 146 | 85 | ||
AK169724.1 | - | 1 - 2 | 150 | 85 | ||
BC027194.1 | - | 1 - 2 | 106 | 4216692 | 85 | |
BC047147.1 | - | 1 - 2 | 171 | 6465983 | 85 | |
BY107376.1 | - | 1 - 2 | 197 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |