Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde4dip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039376.2 | NP_001034465.2 | 27 - 28 | 3980 | 59 | |
NM_001289701.1 | NP_001276630.1 | 27 - 28 | 3980 | 59 | ||
NM_001289702.1 | NP_001276631.1 | 27 - 28 | 3980 | 59 | ||
GenBank mRNA | CB523595.1 | - | 1 - 2 | 445 | 59 | |
CO428791.1 | - | 2 - 3 | 566 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |