Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081081.2 | NP_001074550.1 | 11 - 12 | 1508 | 73 | |
NM_001113383.1 | NP_001106854.1 | 11 - 12 | 1508 | 73 | ||
GenBank mRNA | AK046297.1 | - | 6 - 7 | 430 | 73 | |
AK048072.1 | - | 9 - 10 | 686 | 73 | ||
AK173039.1 | - | 11 - 12 | 1508 | 73 | ||
BC025534.1 | - | 5 - 6 | 292 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |