Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3cd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029837.2 | NP_001025008.2 | 17 - 18 | 2521 | 117 | |
NM_001164049.1 | NP_001157521.1 | 17 - 18 | 2548 | 117 | ||
NM_001164051.1 | NP_001157523.1 | 17 - 18 | 2569 | 117 | ||
NM_001164052.1 | NP_001157524.1 | 15 - 16 | 2263 | 117 | ||
NM_008840.3 | NP_032866.2 | 18 - 19 | 2714 | 117 | ||
GenBank mRNA | AK040867.1 | - | 17 - 18 | 2483 | 117 | |
AK149935.1 | - | 18 - 19 | 2613 | 117 | ||
AK153304.1 | - | 17 - 18 | 2462 | 117 | ||
AK155835.1 | - | 17 - 18 | 2426 | 117 | ||
AK171025.1 | - | 17 - 18 | 2528 | 117 | ||
AK172222.1 | - | 17 - 18 | 2425 | 117 | ||
U86587.1 | - | 15 - 16 | 2231 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |