Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134136.3 | NP_598897.2 | 19 - 20 | 3355 | 74 | |
GenBank mRNA | AK031096.1 | - | 19 - 20 | 3305 | 74 | |
AK031347.1 | - | 19 - 20 | 3286 | 74 | ||
AK089046.1 | - | 19 - 20 | 3324 | 74 | ||
AY267463.1 | - | 19 - 20 | 3285 | 74 | ||
BC056348.1 | - | 19 - 20 | 3307 | 5721407 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |