Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fech | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007998.6 | NP_032024.2 | 2 - 3 | 290 | 117 | |
GenBank mRNA | AK004718.2 | - | 2 - 3 | 291 | 117 | |
AK018738.2 | - | 2 - 3 | 293 | 117 | ||
AK084843.1 | - | 2 - 3 | 188 | 117 | ||
AK159652.1 | - | 2 - 3 | 323 | 117 | ||
BC006746.1 | - | 2 - 3 | 297 | 3256604 | 117 | |
BC094548.1 | - | 2 - 3 | 116 | |||
M59288.1 | - | 1 - 2 | 127 | 117 | ||
M61215.1 | - | 2 - 3 | 283 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |