Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033281.3 | NP_001028453.1 | 2 - 3 | 927 | 68 | |
GenBank mRNA | AK133910.1 | - | 2 - 3 | 928 | 68 | |
BC139295.1 | - | 2 - 3 | 957 | 8862317 | 68 | |
DQ340564.1 | - | 2 - 3 | 503 | 68 | ||
DQ340565.1 | - | 2 - 3 | 422 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |