Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK132691.1 | - | 1 - 2 | 116 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |