Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK132691.1 | - | 2 - 3 | 287 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |