Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080926.1 | NP_001074395.1 | 14 - 15 | 2084 | 64 | |
NR_033496.1 | 15 - 16 | 2462 | 64 | |||
GenBank mRNA | AK030143.1 | - | 11 - 12 | 1594 | 64 | |
AK038580.1 | - | 13 - 14 | 2121 | 64 | ||
BC127061.1 | - | 15 - 16 | 1837 | 64 | ||
D85463.1 | - | 15 - 16 | 2460 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |