Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cc2d1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177045.3 | NP_796019.1 | 2 - 3 | 203 | 57 | |
GenBank mRNA | AK043839.1 | - | 2 - 3 | 204 | 57 | |
AK148162.1 | - | 2 - 3 | 190 | 57 | ||
BC125517.1 | - | 1 - 2 | 93 | 40130032 | 57 | |
BC137743.1 | - | 1 - 2 | 93 | 8860762 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |