Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK029701.1 | - | 5 - 6 | 1070 | 95 | |
AK154663.1 | - | 11 - 12 | 2246 | 95 | ||
BC036324.1 | - | 3 - 4 | 505 | 95 | ||
BC092227.1 | - | 12 - 13 | 2891 | 30531788 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |