Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndfip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022996.1 | NP_075372.1 | 3 - 4 | 421 | 83 | |
GenBank mRNA | AF220209.1 | - | 3 - 4 | 459 | 83 | |
AK050560.1 | - | 3 - 4 | 428 | 83 | ||
BC020359.1 | - | 3 - 4 | 408 | 83 | ||
BC026372.1 | - | 3 - 4 | 421 | 4162070 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |