Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Malt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172833.2 | NP_766421.1 | 1 - 2 | 405 | 139 | |
GenBank mRNA | AK041919.1 | - | 1 - 2 | 410 | 139 | |
AK045200.1 | - | 1 - 2 | 396 | 139 | ||
AK085071.1 | - | 1 - 2 | 405 | 139 | ||
AK164626.1 | - | 1 - 2 | 441 | 139 | ||
AK171981.1 | - | 1 - 2 | 392 | 139 | ||
BC110487.1 | - | 1 - 2 | 379 | 40048907 | 139 | |
BC110488.1 | - | 1 - 2 | 379 | 40048908 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Protease Mammalian Gene Collection Mus musculus 1700 Probe spans exons |