Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159344.1 | NP_001152816.1 | 8 - 9 | 1849 | 94 | |
NM_027195.2 | NP_081471.2 | 8 - 9 | 1849 | 94 | ||
GenBank mRNA | AK138981.1 | - | 3 - 4 | 636 | 94 | |
AK173341.1 | - | 3 - 4 | 639 | 94 | ||
BC075673.1 | - | 8 - 9 | 1851 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |