Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110831.1 | NP_001104301.1 | 3 - 4 | 482 | 64 | |
NM_016878.4 | NP_058574.3 | 3 - 4 | 306 | 64 | ||
GenBank mRNA | AK038646.1 | - | 482 | 64 | ||
AK052001.1 | - | 284 | 64 | |||
AK160084.1 | - | 3 - 4 | 243 | 64 | ||
AK166276.1 | - | 3 - 4 | 256 | 64 | ||
BC092232.1 | - | 3 - 4 | 234 | 30551654 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |