Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177222.4 | NP_796196.3 | 8 - 9 | 846 | 96 | |
GenBank mRNA | AK034148.1 | - | 7 - 8 | 694 | 96 | |
AK132912.1 | - | 8 - 9 | 1070 | 96 | ||
AY423542.1 | - | 8 - 9 | 777 | 96 | ||
AY485607.1 | - | 8 - 9 | 846 | 96 | ||
AY485608.1 | - | 8 - 9 | 846 | 96 | ||
BC072568.1 | - | 6 - 7 | 688 | 96 | ||
BC120743.1 | - | 8 - 9 | 750 | 40129666 | 96 | |
BC125423.1 | - | 8 - 9 | 750 | 40129938 | 96 | |
BC144801.1 | - | 8 - 9 | 750 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |