Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175164.4 | NP_780373.3 | 4 - 5 | 718 | 75 | |
GenBank mRNA | AK129176.1 | - | 4 - 5 | 656 | 75 | |
BC141394.1 | - | 4 - 5 | 587 | 9056007 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |