Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Celf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289613.1 | NP_001276542.1 | 10 - 11 | 1774 | 162 | |
NM_001289616.1 | NP_001276545.1 | 11 - 12 | 1172 | 162 | ||
NM_172434.3 | NP_766022.1 | 10 - 11 | 1687 | 162 | ||
NR_110353.1 | 9 - 10 | 1102 | 162 | |||
GenBank mRNA | AK019578.1 | - | 7 - 8 | 1598 | 162 | |
AK030259.1 | - | 9 - 10 | 1102 | 162 | ||
AK044506.1 | - | 11 - 12 | 1136 | 162 | ||
AY165052.1 | - | 10 - 11 | 1405 | 162 | ||
BC057553.1 | - | 10 - 11 | 1829 | 6401356 | 162 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |