Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neto1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144946.4 | NP_659195.3 | 2 - 3 | 1015 | 115 | |
GenBank mRNA | AF448840.1 | - | 2 - 3 | 348 | 115 | |
AK043710.1 | - | 2 - 3 | 351 | 115 | ||
AK081325.1 | - | 2 - 3 | 1016 | 115 | ||
AY138990.1 | - | 2 - 3 | 84 | 115 | ||
BC051145.1 | - | 2 - 3 | 375 | 6590345 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |