Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyw5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037742.2 | NP_001032831.2 | 6 - 7 | 792 | 103 | |
GenBank mRNA | BC023868.1 | - | 6 - 7 | 784 | 103 | |
BC058590.1 | - | 5 - 6 | 1108 | 103 | ||
BC099495.1 | - | 6 - 7 | 534 | 103 | ||
BC132289.1 | - | 6 - 7 | 641 | 40130566 | 103 | |
BC144766.1 | - | 6 - 7 | 622 | 103 | ||
CF951173.1 | - | 7 - 8 | 568 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |