Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hao2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019545.4 | NP_062418.3 | 7 - 8 | 1129 | 121 | |
GenBank mRNA | AF272947.1 | - | 7 - 8 | 1125 | 121 | |
AJ251820.1 | - | 5 - 6 | 939 | 121 | ||
AK018684.1 | - | 7 - 8 | 1117 | 121 | ||
AK078908.1 | - | 7 - 8 | 1119 | 121 | ||
AK143991.1 | - | 7 - 8 | 1120 | 121 | ||
BC027754.1 | - | 7 - 8 | 1161 | 5064100 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |