Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mboat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083341.1 | NP_001076810.1 | 1 - 2 | 174 | 58 | |
NM_026037.3 | NP_080313.2 | 1 - 2 | 174 | 58 | ||
GenBank mRNA | AB297383.1 | - | 1 - 2 | 73 | 58 | |
AB305046.1 | - | 1 - 2 | 73 | 58 | ||
AK031824.1 | - | 1 - 2 | 159 | 58 | ||
AK076045.1 | - | 1 - 2 | 164 | 58 | ||
AU067514.1 | - | 1 - 2 | 174 | 58 | ||
BC025020.1 | - | 1 - 2 | 124 | 5364068 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |