Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab27b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB067775.1 | - | 1 - 2 | 241 | 115 | |
AF328893.1 | - | 1 - 2 | 254 | 115 | ||
BC018443.1 | - | 1 - 2 | 259 | 3601865 | 115 | |
BC032928.1 | - | 1 - 2 | 174 | 5120275 | 115 | |
BY228140.1 | - | 1 - 2 | 249 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |