Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxxc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028868.3 | NP_083144.1 | 3 - 4 | 488 | 71 | |
GenBank mRNA | AK010337.1 | - | 3 - 4 | 489 | 71 | |
AK083655.1 | - | 3 - 4 | 493 | 71 | ||
AK146688.1 | - | 3 - 4 | 572 | 71 | ||
AY099096.1 | - | 3 - 4 | 460 | 71 | ||
BC030938.1 | - | 3 - 4 | 320 | 3980339 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |