Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ambp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007443.3 | NP_031469.1 | 1 - 2 | 195 | 71 | |
GenBank mRNA | BC021660.1 | - | 1 - 2 | 214 | 4193922 | 71 |
D28812.1 | - | 1 - 2 | 196 | 71 | ||
X68680.1 | - | 1 - 2 | 157 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |