Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400006396.mm.1 |
NM_001113412.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006396.mm.1 |
NM_001113412.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006396.mm.1 |
NM_029347.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006396.mm.1 |
NM_029347.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006396.mm.1 |
TC0400000961.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400006404.mm.1 |
NM_001113412.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006404.mm.1 |
NM_001113412.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006404.mm.1 |
NM_029347.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006404.mm.1 |
NM_029347.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006404.mm.1 |
TC0400000961.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400006407.mm.1 |
NM_001113412.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006407.mm.1 |
NM_001113412.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006407.mm.1 |
NM_029347.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006407.mm.1 |
NM_029347.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006407.mm.1 |
TC0400000961.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400006399.mm.1 |
NM_001113412.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006399.mm.1 |
NM_001113412.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006399.mm.1 |
NM_029347.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006399.mm.1 |
NM_029347.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006399.mm.1 |
TC0400000961.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400006408.mm.1 |
NM_029347.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006408.mm.1 |
NM_029347.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006408.mm.1 |
TC0400000961.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400006400.mm.1 |
NM_001113412.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006400.mm.1 |
NM_001113412.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006400.mm.1 |
NM_029347.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006400.mm.1 |
NM_029347.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006400.mm.1 |
TC0400000961.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400006406.mm.1 |
NM_001113412.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006406.mm.1 |
NM_001113412.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006406.mm.1 |
NM_029347.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006406.mm.1 |
NM_029347.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006406.mm.1 |
TC0400000961.mm.1 |
MTA TC |