Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noc2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021303.2 | NP_067278.2 | 13 - 14 | 1602 | 61 | |
GenBank mRNA | AF155546.1 | - | 1 - 2 | 112 | 61 | |
AK076287.1 | - | 12 - 13 | 1644 | 61 | ||
AK078560.1 | - | 13 - 14 | 1578 | 61 | ||
AK157765.1 | - | 13 - 14 | 1595 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |