Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039201.1 | NP_001034290.1 | 2 - 3 | 224 | 60 | |
NM_001039202.1 | NP_001034291.1 | 2 - 3 | 185 | 60 | ||
NM_029826.2 | NP_084102.1 | 2 - 3 | 185 | 60 | ||
GenBank mRNA | AK002837.1 | - | 2 - 3 | 173 | 60 | |
AK014208.1 | - | 2 - 3 | 179 | 60 | ||
AK137285.1 | - | 2 - 3 | 177 | 60 | ||
AK147794.1 | - | 206 | 60 | |||
AK151285.1 | - | 2 - 3 | 173 | 60 | ||
AK152162.1 | - | 2 - 3 | 173 | 60 | ||
AK152259.1 | - | 2 - 3 | 173 | 60 | ||
BC058177.1 | - | 2 - 3 | 159 | 5250739 | 60 | |
BY069367.1 | - | 2 - 3 | 195 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |