Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183300.2 | NP_899123.2 | 2 - 3 | 171 | 92 | |
GenBank mRNA | DQ303413.1 | - | 2 - 3 | 171 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |