Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kank4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172872.3 | NP_766460.2 | 4 - 5 | 2231 | 86 | |
GenBank mRNA | AK081525.1 | - | 4 - 5 | 2169 | 86 | |
BC060737.1 | - | 4 - 5 | 2245 | 4191502 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |