Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hif3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162950.1 | NP_001156422.1 | 9 - 10 | 1185 | 70 | |
NM_016868.3 | NP_058564.2 | 9 - 10 | 1216 | 70 | ||
GenBank mRNA | AB289606.1 | - | 9 - 10 | 1152 | 70 | |
AF060194.1 | - | 9 - 10 | 1216 | 70 | ||
AK144472.1 | - | 4 - 5 | 559 | 70 | ||
BC120587.1 | - | 9 - 10 | 1169 | 40129510 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |