Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2j9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028979.2 | NP_083255.1 | 3 - 4 | 657 | 112 | |
GenBank mRNA | AF336850.1 | - | 3 - 4 | 657 | 112 | |
AK018422.1 | - | 3 - 4 | 600 | 112 | ||
AK033961.1 | - | 3 - 4 | 627 | 112 | ||
AK037031.1 | - | 3 - 4 | 753 | 112 | ||
BC119556.2 | - | 3 - 4 | 581 | 40057469 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |