Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lix1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163170.1 | NP_001156642.1 | 1 - 2 | 340 | 91 | |
GenBank mRNA | AK051276.1 | - | 1 - 2 | 325 | 91 | |
AK143438.1 | - | 1 - 2 | 341 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |