Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pqlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164420.1 | NP_001157892.1 | 3 - 4 | 635 | 64 | |
NM_001164421.1 | NP_001157893.1 | 3 - 4 | 567 | 64 | ||
NM_001164422.1 | NP_001157894.1 | 3 - 4 | 562 | 64 | ||
GenBank mRNA | AK009256.1 | - | 3 - 4 | 557 | 64 | |
AK153023.1 | - | 3 - 4 | 587 | 64 | ||
AK153176.1 | - | 3 - 4 | 588 | 64 | ||
BC008231.1 | - | 3 - 4 | 504 | 2647794 | 64 | |
BC029179.1 | - | 3 - 4 | 509 | 3487991 | 64 | |
CJ135868.1 | - | 3 - 4 | 587 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |