Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013562.2 | NP_038590.2 | 7 - 8 | 1026 | 98 | |
GenBank mRNA | AK078888.1 | - | 1024 | 98 | ||
AK168682.1 | - | 7 - 8 | 1023 | 98 | ||
AK219989.1 | - | 3 - 4 | 252 | 98 | ||
BC037013.1 | - | 7 - 8 | 1000 | 98 | ||
BC043723.1 | - | 7 - 8 | 993 | 5369100 | 98 | |
X17400.1 | - | 7 - 8 | 1003 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |