Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mllt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027326.3 | NP_081602.3 | 1 - 2 | 328 | 53 | |
GenBank mRNA | AF372456.1 | - | 1 - 2 | 243 | 53 | |
BC021420.1 | - | 1 - 2 | 203 | 5012671 | 53 | |
CJ052430.1 | - | 1 - 2 | 330 | 53 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |