Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc24a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110240.1 | NP_001103710.1 | 2 - 3 | 1270 | 65 | |
GenBank mRNA | BC115867.1 | - | 2 - 3 | 1272 | 40081382 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |