Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efcab7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145549.1 | NP_663524.1 | 1113 | 97 | ||
GenBank mRNA | AK142188.1 | - | 6 - 7 | 1318 | 97 | |
AK143777.1 | - | 1171 | 97 | |||
AK220422.1 | - | 1062 | 97 | |||
BC020077.1 | - | 1113 | 4018470 | 97 | ||
BC112327.1 | - | 6 - 7 | 964 | 5362489 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |