Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aspscr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164224.1 | NP_001157696.1 | 10 - 11 | 1158 | 71 | |
NM_026877.2 | NP_081153.1 | 12 - 13 | 1442 | 71 | ||
NM_198223.2 | NP_937866.1 | 11 - 12 | 1221 | 71 | ||
GenBank mRNA | AK004509.1 | - | 6 - 7 | 454 | 71 | |
AK088197.1 | - | 12 - 13 | 71 | |||
AK146061.1 | - | 10 - 11 | 1159 | 71 | ||
AY349132.1 | - | 10 - 11 | 1211 | 71 | ||
AY349133.1 | - | 12 - 13 | 1359 | 71 | ||
BC019177.1 | - | 12 - 13 | 1385 | 5008127 | 71 | |
BC022115.1 | - | 12 - 13 | 1389 | 5101238 | 71 | |
BC031153.1 | - | 12 - 13 | 1374 | 4976569 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |