Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Satb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163630.1 | NP_001157102.1 | 8 - 9 | 1744 | 111 | |
NM_001163631.1 | NP_001157103.1 | 8 - 9 | 1612 | 111 | ||
NM_001163632.1 | NP_001157104.1 | 8 - 9 | 1585 | 111 | ||
NM_009122.2 | NP_033148.2 | 8 - 9 | 1635 | 111 | ||
GenBank mRNA | AK037740.1 | - | 8 - 9 | 1636 | 111 | |
AK088563.1 | - | 8 - 9 | 1611 | 111 | ||
AK158518.1 | - | 8 - 9 | 1745 | 111 | ||
BC011132.1 | - | 8 - 9 | 1495 | 4164993 | 111 | |
CT010371.1 | - | 7 - 8 | 1423 | 111 | ||
U05252.1 | - | 8 - 9 | 1621 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |